3-Aminobenzenesulfonamide 97+%

Supplier: Thermo Scientific Chemicals
Warning

B21857-06 B21857-14
CAAAB21857-06EA 61.3 CAD
CAAAB21857-06 CAAAB21857-14
3-Aminobenzenesulfonamide 97+%
3-Aminobenzenesulphonamide

Purity: 97+% CAS No.: 98-18-0 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Melting Point: 139-142° MDL No.: MFCD00035781

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00035781
CAS Number: 98-18-0
EINECS: 202-646-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR