1-Iodoperfluorodecane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21845-06
B21845-14
CAAAAB21845-03EA
53.12
CAD
CAAAAB21845-03
CAAAB21845-06
CAAAB21845-14
1-Iodoperfluorodecane 97%
1-Iodoperfluorodecane
Purity: 97%
CAS No.: 423-62-1
Molecular Formula: CF3(CF2)9I
Formula Weight: 645.98
Flash Point: None
Melting Point: 63-67°
Boiling Point: 195-200°
Beilstein Reference: 1810506
MDL No.: MFCD00001065
Formula:
C₁₀F₂₁I Boiling Pt: 195…200 °C Melting Pt: 63…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001065 CAS Number: 423-62-1 EINECS: 207-030-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...