(4-Hydroxyphenyl)acetic acid ethyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl (4-hydroxyphenyl)acetate, Ethyl 2-(4-hydroxyphenyl)acetate

B21832-09 B21832-18
CAAAB21832-09EA 122.61 CAD
CAAAB21832-09 CAAAB21832-18
(4-Hydroxyphenyl)acetic acid ethyl ester 98%
(4-Hydroxyphenyl)acetic acid ethyl ester

Purity: 98% CAS No.: 17138-28-2 Molecular Formula: C10H12O3 Formula Weight: 180.21 Melting Point: 36-38° Boiling Point: 187-189°/20mm MDL No.: MFCD00016491

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 187…189 °C/20 mm
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00016491
CAS Number: 17138-28-2
EINECS: 241-197-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR