2,6-Dimethoxy-p-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dimethoxy-4-hydroxytoluene, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzene, 2,6-Dimethoxy-4-methylphenol

B21801-14 B21801-22
CAAAB21801-14EA 123.71 CAD
CAAAB21801-14 CAAAB21801-22
2,6-Dimethoxy-p-cresol 97%
2,6-Dimethoxy-p-cresol

Purity: 97% CAS No.: 6638-05-7 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 110°(230°F) Odor: Phenol-like Boiling Point: 144-146°/12mm MDL No.: MFCD00017289

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 144…146 °C/12 mm
Melting Pt: 36…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017289
CAS Number: 6638-05-7
EINECS: 229-641-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR