Polyethylene glycol 600
Supplier: Thermo Scientific Chemicals
B21798-0B
B21798-30
CAAAB21798-0BEA
142.52
CAD
CAAAB21798-0B
CAAAB21798-30
Polyethylene glycol 600
Polyethylene glycol 600
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 252°(485°F)
Physical Form: Low-melting solid
Density: 1.120
Refractive Index: 1.4680
Odor: Odorless
pH: 5-7
Melting Point: 17-23°
Merck Reference: 14,7568
MDL No.: MFCD00081839
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ? 250 °C (1013 hPa) Melting Pt: 17…23 °C Density: 1.120 Flash Pt: 252 °C(485 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,7568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...