3-Mercapto-2-pentanone 97% stabilised

Supplier: Thermo Scientific Chemicals

B21797-03 B21797-06
CAAAB21797-03EA 82.48 CAD
CAAAB21797-03 CAAAB21797-06
3-Mercapto-2-pentanone 97% stabilised
3-Mercapto-2-pentanone

Purity: 97% stab. with 0.1% Calcium carbonate CAS No.: 67633-97-0 Molecular Formula: C5H10OS Formula Weight: 118.19 Storage Temperature: -30°C to -10°C Refractive Index: 1.4660 UN No.: 3336 MDL No.: MFCD02093290

Formula: C₅H₁₀OS
MDL Number: MFCD02093290
CAS Number: 67633-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR