(S)-(+)-1,2-Propanediol 97%
Supplier: Thermo Scientific Chemicals
B21773-03
B21773-06
B21773-14
CAAAB21773-03EA
99.86
CAD
CAAAB21773-03
CAAAB21773-06
CAAAB21773-14
(S)-(+)-1,2-Propanediol 97%
(S)-(+)-1,2-Propanediol
Purity: 97%
CAS No.: 4254-15-3
Molecular Formula: C3H8O2
Formula Weight: 76.10
Flash Point: 107°(224°F)
Refractive Index: 1.4320
Boiling Point: 186-188°
Beilstein Reference: 1718871
Merck Reference: 14,7855
Optical Rotation: +16° (neat)
MDL No.: MFCD00004539
Formula:
C₃H₈O₂ Boiling Pt: 186…188 °C Density: 1.040 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004539 CAS Number: 4254-15-3 Merck Index: 14,7855 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...