1-Bromotetrafluoro-2-iodoethane 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
B21771-06
B21771-14
B21771-22
CAAAB21771-06EA
127.03
CAD
CAAAB21771-06
CAAAB21771-14
CAAAB21771-22
1-Bromotetrafluoro-2-iodoethane 97% stabilized
1-Bromotetrafluoro-2-iodoethane
Purity: 97%
CAS No.: 421-70-5
Molecular Formula: BrCF2CF2I
Formula Weight: 306.82
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4330
Boiling Point: 78°
Beilstein Reference: 1688466
MDL No.: MFCD00042136
Formula:
BrF₂CCF₂I Boiling Pt: 78 °C Density: 1.648 Storage Temperature: Ambient |
MDL Number:
MFCD00042136 CAS Number: 421-70-5 EINECS: 207-008-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...