4-Chloro-2-(trifluoromethyl)aniline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Amino-5-chloro-α,α,α-trifluorotoluene, 2-Amino-5-chloro-α,α,α-trifluortoluene, 2-Amino-5-chlorobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-o-toluidine
B21755-14
B21755-22
B21755-36
CAAAB21755-14EA
65.1
CAD
CAAAB21755-14
CAAAB21755-22
CAAAB21755-36
4-Chloro-2-(trifluoromethyl)aniline 97%
4-Chloro-2-(trifluoromethyl)aniline
Purity: 97%
CAS No.: 445-03-4
Molecular Formula: C7H5ClF3N
Formula Weight: 195.57
Flash Point: 95°(203°F)
Refractive Index: 1.5070
Melting Point: 8-9°
Boiling Point: 66-67°/3mm
Beilstein Reference: 2366801
UN No.: 2810
MDL No.: MFCD00007841
Formula:
C₇H₅ClF₃N Boiling Pt: 66…67 °C/3 mm Melting Pt: 8…9 °C Density: 1.386 Flash Pt: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007841 CAS Number: 445-03-4 EINECS: 207-151-3 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...