3,4-Dihydroxybenzhydrazide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dihydroxybenzoic hydrazide, 3,4-Dihydroxybenzoic acid hydrazide, 3,4-Dihydroxybenzene-1-carbohydrazide, 3,4-Dihydroxybenzohydrazide

B21746-06 B21746-14
CAAAB21746-06EA 121.5 CAD
CAAAB21746-06 CAAAB21746-14
3,4-Dihydroxybenzhydrazide 97%
3,4-Dihydroxybenzhydrazide

Purity: 97% CAS No.: 39635-11-5 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: ca 265° dec. Beilstein Reference: 2642968

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Melting Pt: ∼ 265 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00017064
CAS Number: 39635-11-5
EINECS: 254-550-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR