1-Methylcyclohexanecarboxylic acid 99%
Supplier: Thermo Scientific Chemicals
B21713-18
B21713-06
B21713-14
CAAAAB21713-18EA
431.34
CAD
CAAAAB21713-18
CAAAB21713-06
CAAAB21713-14
1-Methylcyclohexanecarboxylic acid 99%
1-Methylcyclohexanecarboxylic acid
Purity: 99%
CAS No.: 1123-25-7
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 100°(212°F)
Melting Point: 36-39°
Boiling Point: 236°
Beilstein Reference: 1859220
MDL No.: MFCD00001463
Formula:
C₈H₁₄O₂ Boiling Pt: 236 °C Melting Pt: 36…39 °C Density: 1.0115 g/cm³ (20 °C) Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001463 CAS Number: 1123-25-7 EINECS: 214-371-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...