2-Bromo-p-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-4-methylphenol, 1-Bromo-2-hydroxy-5-methylbenzene

B21701-06 B21701-14
CAAAB21701-06EA 60.04 CAD
CAAAB21701-06 CAAAB21701-14
2-Bromo-p-cresol 97%
2-Bromo-p-cresol

Purity: 97% CAS No.: 6627-55-0 Molecular Formula: C7H7BrO Formula Weight: 187.04 Flash Point: 110°(230°F) Refractive Index: 1.5780 Odor: Phenol-like Melting Point: 16° Boiling Point: 213-214° MDL No.: MFCD00002151

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 213
Melting Pt: 26
Density: 1.547
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002151
CAS Number: 6627-55-0
EINECS: 229-595-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR