Tris(2-hydroxyethyl)-1,3,5-triazinetrione 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3,5-Tris(2-hydroxyethyl)isocyanuric acid, 1,3,5-Tris(2-hydroxyethyl)cyanuric acid, 1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione, 1,3,5-Tris(2-hydroxyethyl)isocyanurate

B21697-0B B21697-18 B21697-30
CAAAB21697-0BEA 897.44 CAD
CAAAB21697-0B CAAAB21697-18 CAAAB21697-30
Tris(2-hydroxyethyl)-1,3,5-triazinetrione 97%
Tris(2-hydroxyethyl)-1,3,5-triazinetrione

Purity: 97% CAS No.: 839-90-7 Molecular Formula: C9H15N3O6 Formula Weight: 261.24 Melting Point: 136-140° MDL No.: MFCD00003549

Formula: C₉H₁₅N₃O₆
MW: 261.23 g/mol
Boiling Pt: 527
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003549
CAS Number: 839-90-7
EINECS: 212-660-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR