4-Bromo-3,5-xylenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-3,5-dimethylphenol, 2-Bromo-5-hydroxy-3-xylene

B21684-06 B21684-14
CAAAB21684-06EA 86.9 CAD
CAAAB21684-06 CAAAB21684-14
4-Bromo-3,5-xylenol 99%
4-Bromo-3,5-xylenol

Purity: 99% CAS No.: 7463-51-6 Molecular Formula: C8H9BrO Formula Weight: 201.07 Odor: Phenol-like Melting Point: 113-117° Beilstein Reference: 2207127 MDL No.: MFCD00002315

Formula: C₈H₉BrO
MW: 201.06
Boiling Pt: 259
Melting Pt: 116
Storage Temperature: Ambient
MDL Number: MFCD00002315
CAS Number: 7463-51-6
EINECS: 231-255-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR