4-Bromo-2,6-xylenol 99%

Supplier: Thermo Scientific Chemicals
Warning

B21669-36 B21669-14 B21669-22
CAAAAB21669-36EA 545.1 CAD
CAAAAB21669-36 CAAAB21669-14 CAAAB21669-22
4-Bromo-2,6-xylenol 99%
4-Bromo-2,6-xylenol

Purity: 99% CAS No.: 2374-05-2 Molecular Formula: C8H9BrO Formula Weight: 201.07 Odor: Phenol-like Melting Point: 74-78° Beilstein Reference: 1862399 MDL No.: MFCD00002314

Formula: C₈H₉BrO
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00002314
CAS Number: 2374-05-2
EINECS: 219-153-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR