3-(Trifluoromethyl)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21661-03
B21661-06
CAAAAB21661-14EA
957.27
CAD
CAAAAB21661-14
CAAAB21661-03
CAAAB21661-06
3-(Trifluoromethyl)phenylboronic acid 98%
3-(Trifluoromethyl)phenylboronic acid
Purity: 98%
CAS No.: 1423-26-3
Molecular Formula: C7H6BF3O2
Formula Weight: 189.93
Melting Point: 162-166°
Beilstein Reference: 6084746
Formula:
C₇H₆BF₃O₂ Melting Pt: 162…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151854 CAS Number: 1423-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...