2-Chloro-5-methylthiophene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21657-06
B21657-14
CAAAB21657-06EA
135.25
CAD
CAAAB21657-06
CAAAB21657-14
2-Chloro-5-methylthiophene 97%
2-Chloro-5-methylthiophene
Purity: 97%
CAS No.: 17249-82-0
Molecular Formula: C5H5ClS
Formula Weight: 132.61
Flash Point: 47°(117°F)
Refractive Index: 1.5356
Odor: Characteristic
Boiling Point: 155-156°
UN No.: 1992
MDL No.: MFCD01632145
Formula:
C₅H₅ClS Boiling Pt: 155…156 °C Density: 1.21 Flash Pt: 47 °C (117 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01632145 CAS Number: 17249-82-0 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...