10-Methylphenothiazine 98%

Supplier: Thermo Scientific Chemicals

B21648-06 B21648-14
CAAAB21648-06EA 86.74 CAD
CAAAB21648-06 CAAAB21648-14
10-Methylphenothiazine 98%
10-Methylphenothiazine

Purity: 98% CAS No.: 1207-72-3 Molecular Formula: C13H11NS Formula Weight: 213.30 Melting Point: 100-103° MDL No.: MFCD00041836

Formula: C₁₃H₁₁NS
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00041836
CAS Number: 1207-72-3
EINECS: 214-896-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR