4-Iodophenetole 97%

Supplier: Thermo Scientific Chemicals

B21644-06 B21644-14
CAAAB21644-06EA 121.66 CAD
CAAAB21644-06 CAAAB21644-14 CAAAB21644-22
4-Iodophenetole 97%
4-Iodophenetole

Purity: 97% CAS No.: 699-08-1 Molecular Formula: C8H9IO Formula Weight: 248.06 Flash Point: 110°(230°F) Refractive Index: 1.5970 Melting Point: 26-29° Boiling Point: 133-134°/19mm Beilstein Reference: 2042067 MDL No.: MFCD00061124

Formula: C₈H₉IO
Boiling Pt: 133…134 °C/19 mm
Melting Pt: 26…29 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061124
CAS Number: 699-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR