Sodium dicyanoamide 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium dicyanamide

B21634-14 B21634-22 B21634-36
CAAAB21634-14EA 81.69 CAD
CAAAB21634-14 CAAAB21634-22 CAAAB21634-36
Sodium dicyanoamide 96%
Sodium dicyanoamide

Purity: 96% CAS No.: 1934-75-4 Molecular Formula: NaN(CN)2 Formula Weight: 89.03 Melting Point: 300° MDL No.: MFCD00003535

Formula: C₂N₃Na
MW: 89.03 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003535
CAS Number: 1934-75-4
EINECS: 217-703-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR