Chlorophenol red

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide

B21623-06 B21623-14
CAAAB21623-06EA 67.15 CAD
CAAAB21623-06 CAAAB21623-14
Chlorophenol red
Chlorophenol red

CAS No.: 4430-20-0 Molecular Formula: C19H12Cl2O5S Formula Weight: 432.28 Beilstein Reference: 354053 MDL No.: MFCD00005877

Formula: C₁₉H₁₂Cl₂O₅S
MW: 423.27 g/mol
Melting Pt: ∼261 °C
Storage Temperature: Ambient
MDL Number: MFCD00005877
CAS Number: 4430-20-0
EINECS: 224-619-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR