5-Bromo-m-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21610-06
B21610-14
B21610-22
CAAAB21610-06EA
37.92
CAD
CAAAB21610-06
CAAAB21610-14
CAAAB21610-22
5-Bromo-m-xylene 98%
5-Bromo-m-xylene
Purity: 98%
CAS No.: 556-96-7
Molecular Formula: C8H9Br
Formula Weight: 185.07
Flash Point: 87°(188°F)
Refractive Index: 1.5490
Boiling Point: 204°
Beilstein Reference: 1926375
MDL No.: MFCD00000087
Formula:
C₈H₉Br Boiling Pt: 204 °C Density: 1.361 Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000087 CAS Number: 556-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...