1-Iodo-3-nitrobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21605-14
B21605-22
CAAAB21605-14EA
92.59
CAD
CAAAB21605-14
CAAAB21605-22
1-Iodo-3-nitrobenzene 99%
1-Iodo-3-nitrobenzene
Purity: 99%
CAS No.: 645-00-1
Molecular Formula: C6H4INO2
Formula Weight: 249.01
Flash Point: 71°(159°F)
Melting Point: 35-38°
Boiling Point: 279-280°
Beilstein Reference: 1525167
UN No.: 2926
MDL No.: MFCD00007218
Formula:
C₆H₄INO₂ MW: 249.01 g/mol Boiling Pt: 279…280 °C Melting Pt: 35…38 °C Density: 2.227 g/cm³ (20 °C) Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007218 CAS Number: 645-00-1 EINECS: 211-427-9 UN: 2926 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...