Tetrafluoroisophthalonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetrafluoro-1,3-benzenedicarbonitrile, Tetrafluorobenzene-1,3-dicarbonitrile, Tetrafluoro-1,3-dicyanobenzene
B21599-14
B21599-03
CAAAAB21599-14EA
726.8
CAD
CAAAAB21599-14
CAAAB21599-03
Tetrafluoroisophthalonitrile 97%
Tetrafluoroisophthalonitrile
Purity: 97%
CAS No.: 2377-81-3
Molecular Formula: C8F4N2
Formula Weight: 200.10
Melting Point: 76-78°
Beilstein Reference: 6925899
UN No.: 3439
MDL No.: MFCD00013290
Formula:
C₈F₄N₂ MW: 200.1 g/mol Boiling Pt: 283 °C (1013 hPa) Melting Pt: 76…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013290 CAS Number: 2377-81-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...