(2,5-Difluorophenyl)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21595-03
B21595-06
CAAAAB21595-14EA
990.68
CAD
CAAAAB21595-14
CAAAB21595-03
CAAAB21595-06
(2,5-Difluorophenyl)acetic acid 98%
(2,5-Difluorophenyl)acetic acid
Purity: 98%
CAS No.: 85068-27-5
Molecular Formula: C8H6F2O2
Formula Weight: 172.13
Melting Point: 123-127°
Beilstein Reference: 6844956
Formula:
(C₆H₃F₂)CH₂COOH Boiling Pt: 259 °C (1013 hPa) Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010000 CAS Number: 85068-27-5 EINECS: 285-288-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...