2-Chlorobutane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21586-AD
B21586-AK
B21586-AU
CAAAAB21586-ADEA
46.77
CAD
CAAAAB21586-AD
CAAAB21586-AK
CAAAB21586-AU
2-Chlorobutane 98+%
2-Chlorobutane
Purity: 98+%
CAS No.: 78-86-4
Molecular Formula: CH3CHClCH2CH3
Formula Weight: 92.57
Flash Point: -15°(5°F)
Physical Form: Liquid
Density: 0.873
Refractive Index: 1.3970
Odor: Sweet
Melting Point: -141 to -140°
Boiling Point: 67-69°
Beilstein Reference: 1718770
Merck Reference: 14,1561
UN No.: 1127
MDL No.: MFCD00000871
Formula:
C₄H₉Cl MW: 92.57 g/mol Boiling Pt: 67…69 °C Melting Pt: –141 to –140 °C Density: 0.873 Flash Pt: –15 °C (5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000871 CAS Number: 78-86-4 EINECS: 201-151-7 UN: 1127 ADR: 3,II Merck Index: 14,1561 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...