1-Bromo-2,3-difluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Difluorobromobenzene
B21585-03
B21585-06
CAAAB21585-03EA
78.53
CAD
CAAAB21585-03
CAAAB21585-06
1-Bromo-2,3-difluorobenzene 98%
1-Bromo-2,3-difluorobenzene
Purity: 98%
CAS No.: 38573-88-5
Molecular Formula: C6H3BrF2
Formula Weight: 192.99
Flash Point: 61°(141°F)
Refractive Index: 1.5090
Boiling Point: 69-70°/12mm
MDL No.: MFCD00061136
Formula:
C₆H₃BrF₂ MW: 192.99 g/mol Boiling Pt: 69…70 °C/12 mm Density: 1.724 Flash Pt: 61 °C (141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061136 CAS Number: 38573-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...