Pelargonic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21568-AK
B21568-AU
CAAAB21568-AKEA
40.13
CAD
CAAAB21568-AK
CAAAB21568-AU
Pelargonic acid 97%
Pelargonic acid
Purity: 97%
CAS No.: 112-05-0
Molecular Formula: CH3(CH2)7CO2H
Formula Weight: 158.24
Flash Point: 140°(284°F)
Refractive Index: 1.4315
Odor: Unpleasant
pH: 3.5
Melting Point: 10-12°
Boiling Point: 253-255°
Beilstein Reference: 1752351
Merck Reference: 14,7070
UN No.: 3265
MDL No.: MFCD00004433
Formula:
C₉H₁₈O₂ Boiling Pt: 253…255 °C Melting Pt: 10…12 °C Density: 0.905 Flash Pt: 140 °C(284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004433 CAS Number: 112-05-0 EINECS: 203-931-2 UN: 3265 ADR: 8,III Merck Index: 14,7070 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...