Dimethyl-5-hydroxyisophthalate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Hydroxyisophthalic acid dimethyl ester, Dimethyl 5-hydroxyisophthalate
B21565-09
B21565-18
B21565-30
CAAAB21565-09EA
55.3
CAD
CAAAB21565-09
CAAAB21565-18
CAAAB21565-30
Dimethyl-5-hydroxyisophthalate 98%
Dimethyl 5-hydroxyisophthalate
Purity: 98%
CAS No.: 13036-02-7
Molecular Formula: C10H10O5
Formula Weight: 210.19
Melting Point: 162-164°
Beilstein Reference: 2650120
Formula:
C₁₀H₁₀O₅ Melting Pt: 162…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134367 CAS Number: 13036-02-7 EINECS: 235-899-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...