2-Thiopheneethanol 98%

Supplier: Thermo Scientific Chemicals

B21562-06 B21562-14 B21562-22
CAAAB21562-06EA 82.48 CAD
CAAAB21562-06 CAAAB21562-14 CAAAB21562-22
2-Thiopheneethanol 98%
2-Thiopheneethanol

Purity: 98% CAS No.: 5402-55-1 Molecular Formula: C6H8OS Formula Weight: 128.19 Flash Point: 101°(213°F) Refractive Index: 1.5510 Boiling Point: 107-108°/12mm Beilstein Reference: 106985 MDL No.: MFCD00005462

Formula: C₆H₈OS
Boiling Pt: 107…108 °C/12 mm
Density: 1.150
Flash Pt: 101 °C(213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005462
CAS Number: 5402-55-1
EINECS: 226-452-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR