2-Chloro-4-nitrophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chloro-2-hydroxy-5-nitrobenzene

B21561-09 B21561-18 B21561-30
CAAAB21561-09EA 57.67 CAD
CAAAB21561-09 CAAAB21561-18 CAAAB21561-30
2-Chloro-4-nitrophenol 97%
2-Chloro-4-nitrophenol

Purity: 97% CAS No.: 619-08-9 Molecular Formula: C6H4ClNO3 Formula Weight: 173.56 Odor: Characteristic Melting Point: 108-112° Beilstein Reference: 2046372 MDL No.: MFCD00043910

Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00043910
CAS Number: 619-08-9
EINECS: 210-578-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR