(1R)-(-)-Camphor-10-sulfonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid, L(-)-Camphor-10-sulphonic acid, (1R)-(-)-2-Oxobornane-10-sulfonic acid, L(-)-Camphor-10-sulfonic acid, (1R)-(-)-2-Oxobornane-10-sulphonic acid, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (-)-CSA

B21553-36 B21553-14 B21553-22
CAAAAB21553-36EA 766.3 CAD
CAAAAB21553-36 CAAAB21553-14 CAAAB21553-22
(1R)-(-)-Camphor-10-sulfonic acid 98%
(1R)-(-)-Camphor-10-sulphonic acid

Purity: 98+% CAS No.: 35963-20-3 Molecular Formula: C10H16O4S Formula Weight: 232.30 Melting Point: ca 200° Beilstein Reference: 2809676 UN No.: 3261 MDL No.: MFCD00150753

Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: ∼ 200 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150753
CAS Number: 35963-20-3
EINECS: 252-817-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR