(3-Nitrophenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

B21540-03 B21540-06 B21540-14
CAAAB21540-03EA 43.29 CAD
CAAAB21540-03 CAAAB21540-06 CAAAB21540-14
(3-Nitrophenyl)acetic acid 99%
(3-Nitrophenyl)acetic acid

Purity: 99% CAS No.: 1877-73-2 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 117-120° Beilstein Reference: 2050088

Formula: (C₆H₄(NO₂))CH₂COOH
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00007278
CAS Number: 1877-73-2
EINECS: 217-512-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR