Xylene cyanole FF ≥70% (dye content)

Supplier: Thermo Scientific Chemicals
Warning

B21530-09 B21530-18
CAAAB21530-09EA 80.26 CAD
CAAAB21530-09 CAAAB21530-18
Xylene cyanole FF ≥70% (dye content)
Xylene Cyanole FF

A tracking dye for DNA sequencing


CAS No.: 2650-17-1 Molecular Formula: C25H27N2NaO6S2 Formula Weight: 538.61 Melting Point: ca 295° dec. MDL No.: MFCD00019481

Formula: C₂₅H₂₇N₂NaO₆S₂
Melting Pt: ∼ 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040657
CAS Number: 2650-17-1
EINECS: 220-167-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR