(±)-2-Chloromandelic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21525-06
B21525-14
CAAAAB21525-22EA
516.82
CAD
CAAAAB21525-22
CAAAB21525-06
CAAAB21525-14
(±)-2-Chloromandelic acid 98%
(±)-2-Chloromandelic acid
Purity: 98%
CAS No.: 10421-85-9
Molecular Formula: C8H7ClO3
Formula Weight: 186.60
Melting Point: 80-85°
Beilstein Reference: 2367070
Formula:
C₈H₇ClO₃ Boiling Pt: 350 °C (1013 hPa) Melting Pt: 80…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084962 CAS Number: 10421-85-9 EINECS: 233-900-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...