2-Nitrobenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Nitrobenzene-1-sulphonyl chloride, 2-Nitrobenzene-1-sulfonyl chloride
B21522-14
B21522-22
CAAAB21522-14EA
80.74
CAD
CAAAB21522-14
CAAAB21522-22
2-Nitrobenzenesulfonyl chloride 97%
2-Nitrobenzenesulphonyl chloride
Purity: 97%
CAS No.: 1694-92-4
Molecular Formula: C6H4ClNO4S
Formula Weight: 221.62
Melting Point: 64-69°
Beilstein Reference: 521155
UN No.: 3261
MDL No.: MFCD00007430
Formula:
C₆H₄ClNO₄S Melting Pt: 64…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007430 CAS Number: 1694-92-4 EINECS: 216-907-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...