3,3-Dimethyloctane 98%

Supplier: Thermo Scientific Chemicals
Warning

B21520-03 B21520-06
CAAAB21520-03EA 63.99 CAD
CAAAB21520-03 CAAAB21520-06
3,3-Dimethyloctane 98%
3,3-Dimethyloctane

Purity: 98% CAS No.: 4110-44-5 Molecular Formula: C10H22 Formula Weight: 142.29 Boiling Point: 161° UN No.: 3295 MDL No.: MFCD00048874

Formula: C₁₀H₂₂
Boiling Pt: 161 °C
Melting Pt: –69 °C
Density: 0.74
Storage Temperature: Ambient
MDL Number: MFCD00048874
CAS Number: 4110-44-5
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR