4,4,5,5,5-Pentafluoropentan-1-ol 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4,5,5,5-Pentafluoropentanol

B21512-04 B21512-09
CAAAB21512-04EA 57.67 CAD
CAAAB21512-04 CAAAB21512-09
4,4,5,5,5-Pentafluoropentan-1-ol 94%
4,4,5,5,5-Pentafluoropentan-1-ol

Purity: 94% CAS No.: 148043-73-6 Molecular Formula: C5H7F5O Formula Weight: 178.10 Flash Point: 63°(145°F) Refractive Index: 1.3340 Boiling Point: 135° MDL No.: MFCD00153224

Formula: C₅H₇F₅O
MW: 178.1 g/mol
Boiling Pt: 135 °C
Density: 1.350
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00153224
CAS Number: 148043-73-6
EINECS: 421-360-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR