3-Chloro-o-xylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Chloro-2,3-dimethylbenzene, 3-Chloro-1,2-dimethylbenzene
B21505-14
B21505-22
CAAAB21505-14EA
96.06
CAD
CAAAB21505-14
CAAAB21505-22
3-Chloro-o-xylene 97%
3-Chloro-o-xylene
Purity: 97%
CAS No.: 608-23-1
Molecular Formula: C8H9Cl
Formula Weight: 140.61
MDL No.: MFCD00060689
Formula:
C₈H₉Cl MW: 140.61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00060689 CAS Number: 608-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...