Methyl-4-fluorobenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21504-06
B21504-14
B21504-22
CAAAB21504-06EA
90.85
CAD
CAAAB21504-06
CAAAB21504-14
CAAAB21504-22
Methyl-4-fluorobenzoate 97%
Methyl 4-fluorobenzoate
Purity: 97%
CAS No.: 403-33-8
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Flash Point: 77°(170°F)
Refractive Index: 1.4950
Odor: Aromatic
Boiling Point: 195-197°
Beilstein Reference: 2085925
MDL No.: MFCD00017959
Formula:
C₈H₇FO₂ Boiling Pt: 195…197 °C Density: 1.192 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017959 CAS Number: 403-33-8 EINECS: 206-956-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...