2,5-Dichloro-p-phenylenediamine 99%

Supplier: Thermo Scientific Chemicals
Warning

B21494-06 B21494-14
CAAAB21494-06EA 58.93 CAD
CAAAB21494-06 CAAAB21494-14
2,5-Dichloro-p-phenylenediamine 99%
2,5-Dichloro-p-phenylenediamine

Purity: 99% CAS No.: 20103-09-7 Molecular Formula: C6H6Cl2N2 Formula Weight: 177.03 Melting Point: 164-168° MDL No.: MFCD00007902

Formula: C₆H₆Cl₂N₂
Melting Pt: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00007902
CAS Number: 20103-09-7
EINECS: 243-512-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR