10-Bromo-1-decanol 95%

Supplier: Thermo Scientific Chemicals
Warning

B21483-03 B21483-06
CAAAB21483-03EA 45.35 CAD
CAAAB21483-03 CAAAB21483-06
10-Bromo-1-decanol 95%
10-Bromo-1-decanol

Purity: 95% CAS No.: 53463-68-6 Molecular Formula: Br(CH2)10OH Formula Weight: 237.19 Flash Point: 110°(230°F) Refractive Index: 1.4760 Boiling Point: 165-169°/10mm Beilstein Reference: 1738593 MDL No.: MFCD00041681

Formula: HO(CH₂)₉CH₂Br
Boiling Pt: 165…169 °C/10 mm
Density: 1.190
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041681
CAS Number: 53463-68-6
EINECS: 258-572-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR