2,5-Dimethylstyrene 97% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
B21476-03
B21476-06
B21476-14
CAAAB21476-03EA
51.51
CAD
CAAAB21476-03
CAAAB21476-06
CAAAB21476-14
2,5-Dimethylstyrene 97% stabilised
2,5-Dimethylstyrene
Purity: 97%
CAS No.: 2039-89-6
Molecular Formula: C10H12
Formula Weight: 132.21
Storage Temperature: 2°C to 8°C
Flash Point: 63°(145°F)
Refractive Index: 1.5390
Melting Point: -35°
Boiling Point: 83-84°/24mm
Beilstein Reference: 2203656
MDL No.: MFCD00008614
Formula:
C₁₀H₁₂ MW: 132.21 Boiling Pt: 83…84 °C/24 mm Melting Pt: –35 °C Density: 0.904 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008614 CAS Number: 2039-89-6 EINECS: 218-028-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...