L(+)-Asparagine, anhydrous 99%
Supplier: Thermo Scientific Chemicals
B21473-0E
B21473-22
B21473-36
CAAAB21473-0EEA
1273.48
CAD
CAAAB21473-0E
CAAAB21473-22
CAAAB21473-36
L(+)-Asparagine, anhydrous 99%
L(+)-Asparagine
Purity: 99%
CAS No.: 70-47-3
Molecular Formula: C4H8N2O3
Formula Weight: 132.12
Melting Point: 234-236° dec.
Beilstein Reference: 1723527
Merck Reference: 14,837
MDL No.: MFCD00064401
Formula:
H₂NCOCH₂CH(NH₂)CO₂H Boiling Pt: 343 °C (1013 hPa) Melting Pt: 234…236 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064401 CAS Number: 70-47-3 EINECS: 200-735-9 Merck Index: 14,837 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...