4,4'-Di-tert-butylbiphenyl 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-tert-butyl-4-(4-tert-butylphenyl)benzene, 4,4'-Di-tert-butyl-1,1'-biphenyl

B21470-22 B21470-06 B21470-14
CAAAAB21470-22EA 872.16 CAD
CAAAAB21470-22 CAAAB21470-06 CAAAB21470-14
4,4'-Di-tert-butylbiphenyl 99%
4,4'-Di-tert-butylbiphenyl

Purity: 99% CAS No.: 1625-91-8 Molecular Formula: C20H26 Formula Weight: 266.43 Melting Point: 126-129° Boiling Point: 190-192°/13mm MDL No.: MFCD00008834

Formula: C₂₀H₂₆
MW: 266.43 g/mol
Boiling Pt: 228
Melting Pt: 127
Storage Temperature: Ambient
MDL Number: MFCD00008834
CAS Number: 1625-91-8
EINECS: 216-615-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR