2,6-Dihydroxynaphthalene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21465-06
B21465-14
B21465-22
CAAAB21465-06EA
115.97
CAD
CAAAB21465-06
CAAAB21465-14
CAAAB21465-22
2,6-Dihydroxynaphthalene 98%
2,6-Dihydroxynaphthalene
Purity: 98%
CAS No.: 581-43-1
Molecular Formula: C10H8O2
Formula Weight: 160.17
Melting Point: 223-225°
Beilstein Reference: 1238082
Formula:
C₁₀H₈O₂ Boiling Pt: 375 °C (1013 hPa) Melting Pt: 223…225 °C Density: 1.33 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004082 CAS Number: 581-43-1 EINECS: 209-465-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...