1,2-Epoxyoctan 97%

Supplier: Thermo Scientific Chemicals
Warning

B21463-14 B21463-22
CAAAB21463-14EA 224.36 CAD
CAAAB21463-14 CAAAB21463-22
1,2-Epoxyoctan 97%
1,2-Epoxyoctan

Purity: 97% CAS No.: 2984-50-1 Molecular Formula: C8H16O Formula Weight: 128.22 Storage Temperature: 2°C to 8°C Flash Point: 37°(98°F) Refractive Index: 1.4200 Boiling Point: 167° Beilstein Reference: 79912 UN No.: 1993 MDL No.: MFCD00005157

Formula: C₈H₁₆O
Boiling Pt: 167 °C
Density: 0.838
Flash Pt: 37 °C (98 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005157
CAS Number: 2984-50-1
EINECS: 221-047-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR