4,4,4-Trifluoro-1-butanol 97%

Supplier: Thermo Scientific Chemicals
Warning

B21460-03 B21460-06
CAAAB21460-03EA 115.81 CAD
CAAAB21460-03 CAAAB21460-06
4,4,4-Trifluoro-1-butanol 97%
4,4,4-Trifluoro-1-butanol

Purity: 97% CAS No.: 461-18-7 Molecular Formula: CF3(CH2)3OH Formula Weight: 128.09 Flash Point: 52°(125°F) Refractive Index: 1.3430 Boiling Point: 122-124° Beilstein Reference: 1736068 UN No.: 1993 MDL No.: MFCD00041483

Formula: C₄H₇F₃O
MW: 128.09
Boiling Pt: 122
Density: 1.193
Flash Pt: 52 °C (125 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041483
CAS Number: 461-18-7
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR