5-Fluoroanthranilic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21458-14
B21458-03
B21458-06
CAAAAB21458-14EA
1252.94
CAD
CAAAAB21458-14
CAAAB21458-03
CAAAB21458-06
5-Fluoroanthranilic acid 98+%
5-Fluoroanthranilic acid
Purity: 98+%
CAS No.: 446-08-2
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Melting Point: 182-184°
Beilstein Reference: 2803664
Formula:
C₇H₆FNO₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 182…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055566 CAS Number: 446-08-2 EINECS: 207-159-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...