3-Methylanisole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21455-18
B21455-30
CAAAB21455-18EA
63.36
CAD
CAAAB21455-18
CAAAB21455-30
3-Methylanisole 99%
3-Methylanisole
Purity: 99%
CAS No.: 100-84-5
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 54°(129°F)
Refractive Index: 1.5130
Odor: Pleasant
Boiling Point: 175-176°
Beilstein Reference: 907023
UN No.: 3271
MDL No.: MFCD00008395
Formula:
C₈H₁₀O Boiling Pt: 175…176 °C Melting Pt: –47 °C Density: 0.969 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008395 CAS Number: 100-84-5 EINECS: 202-893-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...